Platinum 3% on activated carbon wetted with ca. 50% water, powder, reduced

Supplier: Thermo Scientific Chemicals

038316-06 038316-14
AA38316-06EA 73.2 USD
AA38316-06 AA38316-14 AA38316-22
Platinum 3% on activated carbon wetted with ca. 50% water, powder, reduced
Platinum

Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 7440-06-4 Physical Form: 900-1100 m²/g MDL No.: MFCD00011179


Ordering information: Sold on a dry weight basis. Unit weight excludes water weight

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR