Platinum 1% on activated carbon wetted with ca. 50% water, powder, reduced

Supplier: Thermo Scientific Chemicals

038312-14
AA38312-22EA 434.01 USD
AA38312-22 AA38312-14
Platinum 1% on activated carbon wetted with ca. 50% water, powder, reduced
Platinum

Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 7440-06-4 Physical Form: 900-1100 m²/g MDL No.: MFCD00011179


Ordering information: Sold on a dry weight basis. Unit weight excludes water weight

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR