Glassy carbon, plate, Thickness 1 mm (0.04 in) Type 1

Supplier: Thermo Scientific Chemicals

038024-FI 038024-GH
AA38024-FIEA 331.27 USD
AA38024-FI AA38024-GH
Glassy carbon, plate, Thickness 1 mm (0.04 in) Type 1
Glassy carbon
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR