Glassy carbon, plate, Thickness 2 mm (0.08 in) Type2

Supplier: Thermo Scientific Chemicals

038022-FI 038022-GH
AA38022-FIEA 329 USD
AA38022-FI AA38022-GH
Glassy carbon, plate, Thickness 2 mm (0.08 in) Type2
Glassy carbon
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR