Glassy carbon, spherical powder 0,4-12 µm Type 1

Supplier: Thermo Scientific Chemicals
Warning

038004-30 038004-18 038004-09
AA38004-30EA 775 USD
AA38004-30 AA38004-18 AA38004-09
Glassy carbon, spherical powder 0,4-12 µm Type 1
Glassy carbon

Appearance: Black UN No.: 1325 MDL No.: MFCD00133992

Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
UN: 1325
ADR: 4.1 III
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR