Mercury(II) thiocyanate
Supplier: Thermo Scientific Chemicals
MDL: MFCD00011039
Beilstein Registry No.: 3561344
CAS No.: 592-85-8
Molecular Formula: C2HgN2S2
Formula Weight: 316.75
Physical Form: Powder
Density: 3.71
Sensitivity: Hygroscopic
Appearance: White to pale yellow
Odor: Odorless
Melting Point: ca 165° dec.
Beilstein Reference: 3561344
Merck Reference: 14,5890
UN No.: 1646
MDL No.: MFCD00011039
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...