Methyl cellulose, 4000 mPa.S (2% H₂O), powder
Supplier: Thermo Scientific Chemicals
Synonyms:
Cellulose methyl ether
036718-22
036718-36
036718-A4
AA36718-22EA
63.4
USD
AA36718-22
AA36718-36
AA36718-A4
Methyl cellulose, 4000 mPa.S (2% H₂O), powder
Methyl cellulose
MDL: MFCD00081763
Notes: Viscosity 4000 cPs for a 2% aqueous solution at 20°C
CAS No.: 9004-67-5
Physical Form: Powder
Odor: Odorless
Merck Reference: 14,6040
MDL No.: MFCD00081763
Caution: Incompatible with strong oxidizing agents, bleach, perchloric acid, nitric acid, perchlorates, alkali nitrates, alkali nitrites and calcium oxide.
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 Merck Index: 14,6040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...