Cyclohexyldiphenylphosphine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Diphenylcyclohexylphosphine

036708-06 036708-03 036708-22
AA36708-14EA 323.68 USD
AA36708-14 AA36708-06 AA36708-03 AA36708-22
Cyclohexyldiphenylphosphine 98%
Cyclohexyldiphenylphosphine

Purity: 98% CAS No.: 6372-42-5 Molecular Formula: C18H21P Formula Weight: 268.34 Flash Point: 110°(230°F) Physical Form: Solid Sensitivity: Air Sensitive Melting Point: 58-62° MDL No.: MFCD00046360

Formula: C₁₈H₂₁P
MW: 268.33 g/mol
Melting Pt: 58…62 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00046360
CAS Number: 6372-42-5
EINECS: 228-904-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR