Cyclohexyldiphenylphosphine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Diphenylcyclohexylphosphine
036708-06
036708-03
036708-22
AA36708-14EA
323.68
USD
AA36708-14
AA36708-06
AA36708-03
AA36708-22
Cyclohexyldiphenylphosphine 98%
Cyclohexyldiphenylphosphine
Purity: 98%
CAS No.: 6372-42-5
Molecular Formula: C18H21P
Formula Weight: 268.34
Flash Point:
Formula:
C₁₈H₂₁P MW: 268.33 g/mol Melting Pt: 58…62 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00046360 CAS Number: 6372-42-5 EINECS: 228-904-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...