Hydrobromic acid 47.0 - 49.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00011323
Beilstein Registry No.: 3587158
Fieser: 1,450 2,214 3,154 18,180
CAS No.: 10035-10-6
Molecular Formula: HBr
Formula Weight: 80.92
Physical Form: Liquid
Density: 1.490
Sensitivity: Air Sensitive
Appearance: Colorless to yellow
Beilstein Reference: 3587158
Merck Reference: 14,4778
UN No.: 1788
MDL No.: MFCD00011323
Formula:
HBr Density: 1.490 Storage Temperature: Ambient |
MDL Number:
MFCD00011323 CAS Number: 10035-10-6 EINECS: 233-113-0 UN: 1788 ADR: 8,II Merck Index: 14,4778 |
Specification Test Results
Form | Liquid |
Cl | 0.05% |
I | 0.003% |
PO3 | 0.001% |
Sulfate and sulfite (as SO3) | 0.003% |
Heavy Metals (as Pb) | 5 ppm |
Fe | 1 ppm |
Se | 0.01 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...