Molybdenum(VI) oxide ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
Source of molybdenum compounds, in agriculture, in analytical chemistry, as a corrosion inhibitor, in ceramic glazes, enamels and pigments, as a catalyst
Purity: 99.5% min
CAS No.: 1313-27-5
Molecular Formula: MoO3
Formula Weight: 143.94
Physical Form: Powder
Density: 4.692
Appearance: White to grey
Odor: Odorless
Melting Point: 795°
Boiling Point: 1155° subl.
Solubility: Slightly soluble in water. Soluble in concentrated mineral acids, alkali hydroxide solutions
Merck Reference: 14,6239
UN No.: 3288
MDL No.: MFCD00003469
Source of molybdenum compounds, in agriculture, in analytical chemistry, as a corrosion inhibitor, in ceramic glazes, enamels and pigments, as a catalyst
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...