Molybdenum(VI) oxide ≥99.5% ACS

Supplier: Thermo Scientific Chemicals
Danger

036687-22 036687-36
AA36687-22EA 73.3 USD
AA36687-22 AA36687-36
Molybdenum(VI) oxide ≥99.5% ACS
Formula: MoO₃
Boiling Pt: 1155 °C (subl.)
Melting Pt: 795 °C
Density: 4.692
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 14,6239

Order Now

Specification Test Results

Insoluble in dilute ammonium hydroxide 0.01%
Cl 0.002%
NO4 Pass test (limit about 0.003%)
Arsenate phosphate and silicate (as SiO2) 0.00001
PO3 5%
SO3 0.02%
NH5 0.002%
Heavy metals (as Pb) 0.005%%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR