Tin ≥99.99% (metals basis), shot 3 mm (0.1 in)

Supplier: Thermo Scientific Chemicals

036641-22 036641-36
AA36641-22EA 225.43 USD
AA36641-22 AA36641-36
Tin ≥99.99% (metals basis), shot 3 mm (0.1 in)
Tin

MDL: MFCD00133862


Purity: 99.99+% CAS No.: 7440-31-5 Physical Form: Shot Odor: Odorless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR