Strontium carbonate ≥99.994% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

036618-22 036618-36 036618-14
AA36618-22EA 498 USD
AA36618-22 AA36618-36 AA36618-14
Strontium carbonate ≥99.994% (metals basis), Puratronic®
Strontium carbonate

Powder, 22 mesh. Purity based on metal contaminates only.


Purity: 99.994% CAS No.: 1633-05-2 Molecular Formula: SrCO3 Formula Weight: 147.63 Physical Form: -22 Mesh Powder Density: 3.70 Melting Point: 1100° dec. Merck Reference: 14,8838 MDL No.: MFCD00011250

Formula: CO₃Sr
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1100 °C (dec.)
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,8838

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR