Formic acid ≥96% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00003297
Beilstein Registry No.: 1209246
Fieser: 1,404 8,232 9,226 11,243 13,137 18,163 19,148 20,168 21,193
Purity: 96+%
CAS No.: 64-18-6
Molecular Formula: CH2O2
Formula Weight: 46.03
Flash Point: 50°(156°F)
Physical Form: Liquid
Density: 1.220
Refractive Index: 1.3710
Appearance: Colorless
Melting Point: 7-9°
Boiling Point: 100-101°
Beilstein Reference: 1209246
Merck Reference: 14,4241
UN No.: 1779
MDL No.: MFCD00003297
Formula:
HCO₂H Boiling Pt: 100…101 °C Melting Pt: 7…9 °C Density: 1.220 Flash Pt: 50 °C (156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003297 CAS Number: 64-18-6 EINECS: 200-579-1 UN: 1779 ADR: 8,II Merck Index: 14,4241 |
|
Specification Test Results
Form | Liquid |
Dilution test | Pass test |
Evaporation residue | 0.003% |
CH3COOH | 0.4% |
NH3 | 0.005% |
Cl | 0.001% |
SO3 | Pass test |
Heavy Metals (as Pb) | 0.001% |
Fe | 0.001% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...