Methanol-[D₄] (99.8% D)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methanol-[²H₄], Hydroxy methane-D4, Methanol-d4, Methyl-D3 alcohol-D, Carbinol-D4, Wood alcohol-D4, Methyl alcohol-D4, Tetradeuteromethanol
036501-03
036501-14
036501-06
AA36501-03EA
63.48
USD
AA36501-03
AA36501-14
AA36501-06
Methanol-[D₄] (99.8% D)
Methanol-[D4]
Purity: 99.8%
CAS No.: 811-98-3
Molecular Formula: CD3OD
Formula Weight: 36.07
Physical Form: Liquid
Density: 0.89
Melting Point: -98°
Boiling Point: 64.7°
UN No.: 1230
MDL No.: MFCD00044637
Formula:
D₃COD MW: 36.01 g/mol Boiling Pt: 65.3 Melting Pt: -99 Density: 0.89 Flash Pt: 11 Storage Temperature: Ambient |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 EINECS: 212-378-6 UN: 1230 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...