Sodium tetraphenylborate ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Tetraphenyl sodium borate, Sodium tetraphenylboron, Tetraphenylboron sodium salt
036430-04
036430-09
036430-18
AA36430-04EA
28.46
USD
AA36430-04
AA36430-09
AA36430-18
Sodium tetraphenylborate ≥99.5% ACS
Sodium tetraphenylborate
MDL: MFCD00011494
Beilstein Registry No.: 3599783
Purity: 99.5% min
CAS No.: 143-66-8
Molecular Formula: C24H20BNa
Formula Weight: 342.23
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
pH: 8
Melting Point:
Formula:
(C₆H₅)₄BNa Melting Pt: >300 °C Density: 1.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011494 CAS Number: 143-66-8 EINECS: 205-605-5 UN: 2811 ADR: 6.1,III Merck Index: 14,8690 |
Specification Test Results
Loss on drying at 150°C | 0.5% |
Clarity of solution | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...