Silicon dioxide ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

036346-22 036346-14
AA36346-22EA 142 USD
AA36346-22 AA36346-14
Silicon dioxide ≥99.99% (metals basis)
Silicium dioxide

MDL: MFCD00011232 Notes: For vacuum deposition


Purity: 99.99% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: 3-12mm Fused Lump Density: 2.2-2.6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Density: 2.2…2.6
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,8493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR