Indium phosphide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Indium(III) phosphide, Indium phosphide

036283-03 036283-06
AA36283-03EA 204 USD
AA36283-03 AA36283-06
Indium phosphide ≥99.999% (metals basis)
Indium phosphide

MDL: MFCD00016153


Purity: 99.999% CAS No.: 22398-80-7 Molecular Formula: InP Formula Weight: 145.79 Physical Form: 6mm & Down Density: 4.81 Melting Point: 1070° Merck Reference: 14,4953 MDL No.: MFCD00016153

Formula: InP
Melting Pt: 1070 °C
Density: 4.81
Storage Temperature: Ambient
MDL Number: MFCD00016153
CAS Number: 22398-80-7
EINECS: 244-959-5
Merck Index: 14,4953

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR