Antimony(III) telluride ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
036282-18
036282-09
AA36282-18EA
577
USD
AA36282-18
AA36282-09
Antimony(III) telluride ≥99.999% (metals basis)
Antimony (III) telluride
For vacuum deposition
Purity: 99.999%
CAS No.: 1327-50-0
Molecular Formula: Sb2Te3
Formula Weight: 626.30
Physical Form: 3-12mm Lump
Density: 6.5
Appearance: Grey
Odor: Odorless
Melting Point: 629°
UN No.: 1549
MDL No.: MFCD00016320
For vacuum deposition
Formula:
Sb₂Te₃ Melting Pt: 629 °C Density: 6.5 Storage Temperature: Ambient |
MDL Number:
MFCD00016320 CAS Number: 1327-50-0 EINECS: 215-480-9 UN: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...