Hexachloroplatinic(IV) acid hexahydrate ≥99.95% (metals basis) Pt ≥ 37.5%, Premion® ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate

036259-03 036259-06 036259-14
AA36259-03EA 173 USD
AA36259-03 AA36259-06 AA36259-14
Hexachloroplatinic(IV) acid hexahydrate ≥99.95% (metals basis) Pt ≥ 37.5%, Premion® ACS
Hexachloroplatinic (IV) acid hexahydrate

MDL: MFCD00149910 Fieser: 1890 487 13145 15135


Purity: 99.95% CAS No.: 18497-13-7 Molecular Formula: H2Cl6Pt.6H2O Formula Weight: 517.91 (409.82anhy) Physical Form: Lump Density: 2.430 Appearance: Red-brown Odor: Odorless Melting Point: 60° dec. Merck Reference: 14,7526 UN No.: 2507 MDL No.: MFCD00149910

Formula: H₂PtCl₆·6H₂O
Melting Pt: 60 °C (dec.)
Density: 2.430
Storage Temperature: Ambient
MDL Number: MFCD00149910
CAS Number: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8,III
Merck Index: 14,7526

Order Now

Specification Test Results

Solubility in alcohol Pass Test
Alkali and other salts (as sulfates) 0.05%
Suitability for potassium determinations Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR