Ammonium thiocyanate ≥97.5% ACS

Supplier: Thermo Scientific Chemicals
Warning

036247-30 036247-A1
AA36247-30EA 98.05 USD
AA36247-30 AA36247-A1
Ammonium thiocyanate ≥97.5% ACS
Ammonium thiocyanate

Ammonium thiocyanate has many uses in manufacturing chemicals such as metal thiocyanates [CuSCN, Ca(SCN)2, NaSCN, KSCN], and in a number of pharmaceuticals. It is used in freezing solutions, pickling, printing, and corrosion inhibitor against acid gases. It is used in photography industry as a stabilizer or accelerator. Used in fertilizers, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe.


Purity: 97.5% min CAS No.: 1762-95-4 Molecular Formula: NH4SCN Formula Weight: 76.12 Physical Form: Crystalline Density: 1.305 Melting Point: 147-149° Solubility: Soluble in water, ethanol, methanol, ammonia, and acetone. Practically insoluble in CHCl3 and ethyl acetate. Beilstein Reference: 3595135 Merck Reference: 14,561 MDL No.: MFCD00011428


Fertilizers, photography, fabric dyeing, zinc coating, electroplating, and as a separator of Zr and Hf, also Au and Fe


Solubility: Soluble in water, ethanol, methanol, ammonia, and acetone. Practically insoluble in CHCl3 and ethyl acetate.

Formula: NH₄SCN
Melting Pt: 147…149 °C
Density: 1.305
Storage Temperature: Ambient
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 14,561

Order Now

Specification Test Results

pH of a 5% solution 4.5-6.0 at 25°
Insoluble matter 0.005%
Residue after ignition 0.025%
Cl 0.005%
SO3 0.005%
Heavy Metals (as Pb) 5 ppm
Fe 3 ppm
Iodine-consuming substances 0.004 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR