Yttrium aluminum oxide ≥99.99%
Supplier: Thermo Scientific Chemicals
036241-14
036241-22
AA36241-14EA
123
USD
AA36241-14
AA36241-22
Yttrium aluminum oxide ≥99.99%
Yttrium aluminum oxide
Applications: Can be used as a sputtering target to prepare bubble memory devices.
Purity: 99.99%
CAS No.: 12005-21-9
Molecular Formula: Y3Al5O12
Formula Weight: 593.62
Physical Form: 3-12mm Fused Lump
Density: 4.65
Refractive Index: 1.833
Melting Point: 1760°
MDL No.: MFCD00213958
Applications: Can be used as a sputtering target to prepare bubble memory devices.
Formula:
Y₃Al₅O₁₂ Melting Pt: 1760 °C Density: 4.65 Storage Temperature: Ambient |
MDL Number:
MFCD00213958 CAS Number: 12005-21-9 EINECS: 234-465-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...