Benzoic acid ≥99.5% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
036230-36
036230-14
036230-22
AA36230-36EA
165.85
USD
AA36230-36
AA36230-14
AA36230-22
Benzoic acid ≥99.5% ACS
Benzoic acid
MDL: MFCD00002398
Beilstein Registry No.: 636131
Fieser: 1,49
Purity: 99.5% min
CAS No.: 65-85-0
Molecular Formula: C7H6O2
Formula Weight: 122.12
Flash Point: 121°(250°F)
Physical Form: Crystalline
Density: 1.32
Appearance: White
Odor: Weak, characteristic
pH: 2.5-3.5
Melting Point: 121-124°
Boiling Point: 249°
Beilstein Reference: 636131
Merck Reference: 14,1091
MDL No.: MFCD00002398
Formula:
C₇H₆O₂ Boiling Pt: 249 °C Melting Pt: 121…124 °C Density: 1.32 Flash Pt: 121 °C (250 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 EINECS: 200-618-2 REACH: 01-2119455536-33 Merck Index: 14,1091 |
|
Specification Test Results
Freezing point | 122°-123° |
Residue after ignition | 0.005% |
Insoluble in methanol | 0.005 AU |
Chlorine compounds (as Cl) | 0.005% |
Sulfur compounds (as S) | 0.002% |
Heavy Metals (as Pb) | 5 ppm |
Substances reducing permanganate | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...