Manganese ≥99.95% (metals basis), irregular pieces

Supplier: Thermo Scientific Chemicals

036221-14 036221-22
AA36221-14EA 117.75 USD
AA36221-14 AA36221-22
Manganese ≥99.95% (metals basis), irregular pieces
Manganese

For vacuum deposition


Purity: 99.95% CAS No.: 7439-96-5 Physical Form: Pieces Odor: Odorless MDL No.: MFCD00011111


For vacuum deposition

Formula: Mn
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR