Diiron arsenide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron arsenide

036190-09
AA36190-18EA 1524.03 USD
AA36190-18 AA36190-09
Diiron arsenide ≥99.5% (metals basis)
Diiron arsenide

MDL: MFCD00151533


Purity: 99.5% CAS No.: 12005-88-8 Molecular Formula: Fe2As Formula Weight: 186.62 Physical Form: 10mm & down lump Odor: Odorless UN No.: 1557 MDL No.: MFCD00151533

Formula: AsFe₂
Storage Temperature: Ambient
MDL Number: MFCD00151533
CAS Number: 12005-88-8
EINECS: 234-474-7
UN: 1557
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR