Zirconium Crucibles with Snap-On Ring
Supplier: Thermo Scientific Chemicals
036020-KT
AA36020-KTEA 268 USD
AA36020-KT
Zirconium Crucibles with Snap-On Ring
Crucibles
Zirconium's distinctive properties make it a popular choice for fusions and fluxes in analytical chemistry and chemical processing.
  • Crucibles feature a snap-on ring
  • Removable ring snap-fits into a machined groove approximately 0.20" below the top of the crucible
Fluxes that can be used in these zirconium crucibles include sodium peroxide fusion, sodium carbonate fusion, and lithium salt fusion. Prolonged exposure to air at temperatures of more than 750 °C can have a negative effect on zirconium.