Bismuth ≥99.999% (metals basis), rods, Diameter 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

035854-22 035854-14
AA35854-22EA 1018 USD
AA35854-22 AA35854-14
Bismuth ≥99.999% (metals basis), rods, Diameter 12.7 mm (0.5 in)
Bismuth

MDL: MFCD00134033


Purity: 99.999+% CAS No.: 7440-69-9 Physical Form: ≈12.5g/cm Odor: Odorless MDL No.: MFCD00134033

Formula: Bi
Boiling Pt: 1490 °C (1013 hPa)
Melting Pt: 271…272 °C
Density: 9.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134033
CAS Number: 7440-69-9
EINECS: 231-177-4
Merck Index: 12,01297

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR