Boron tribromide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
035841-09
AA35841-09EA
199
USD
AA35841-09
AA35841-18
Boron tribromide ≥99.999% (metals basis)
Boron tribromide
MDL: MFCD00011312
Purity: 99.999%
CAS No.: 10294-33-4
Molecular Formula: BBr3
Formula Weight: 250.52
Physical Form: Liquid
Density: 2.643
Appearance: Light yellow
Odor: Acrid
Melting Point: -46°
Boiling Point: 90°
Merck Reference: 14,1347
UN No.: 2692
MDL No.: MFCD00011312
Formula:
BBr₃ MW: 250.52 g/mol Boiling Pt: 90 °C Melting Pt: –46 °C Density: 2.643 Storage Temperature: Ambient |
MDL Number:
MFCD00011312 CAS Number: 10294-33-4 EINECS: 233-657-9 UN: 2692 ADR: 8,I Merck Index: 14,1347 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...