Strontium carbonate ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals

035793-14 035793-22 035793-36
AA35793-14EA 127.59 USD
AA35793-14 AA35793-22 AA35793-36
Strontium carbonate ≥99.99% (metals basis)
Strontium carbonate

MDL: MFCD00011250 Notes: 1340°C -CO2


Purity: 99.99% CAS No.: 1633-05-2 Molecular Formula: SrCO3 Formula Weight: 147.63 Physical Form: Powder Density: 3.70 Melting Point: 1100° dec. Merck Reference: 14,8838 MDL No.: MFCD00011250

Formula: CO₃Sr
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1100 °C (dec.)
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,8838

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR