Holmium, Plasma Standard Solution, Specpure®, Ho 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
035756-AE
AA35756-AEEA
83.8
USD
AA35756-AE
AA35756-AP
Holmium, Plasma Standard Solution, Specpure®, Ho 1000 µg/ml, Thermo Scientific
Standards
Cationic Standards
Holmium Standards
Molecular Formula: Ho2O3 in 5% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011049
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...