Chromium(III) fluoride ≥98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Chromium trifluoride
035741-04
035741-09
AA35741-04EA
164
USD
AA35741-04
AA35741-09
Chromium(III) fluoride ≥98% (metals basis)
Chromium (III) fluoride
MDL: MFCD00016045
Purity: 99.98%
CAS No.: 7788-97-8
Molecular Formula: CrF3
Formula Weight: 108.99
Physical Form: Powder
Density: 3.8
Appearance: Dark green
Odor: Odorless
Melting Point:
Formula:
CrF₃ Melting Pt: >1000 °C Density: 3.8 Storage Temperature: Ambient |
MDL Number:
MFCD00016045 CAS Number: 7788-97-8 EINECS: 232-137-9 UN: 1756 ADR: 8,II Merck Index: 14,2223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...