Strontium chloride, ultra dry ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
035719-06
035719-14
AA35719-06EA
103.93
USD
AA35719-06
AA35719-14
Strontium chloride, ultra dry ≥99.995% (metals basis)
Strontium chloride
MDL: MFCD00011249
Purity: 99.995%
CAS No.: 10476-85-4
Molecular Formula: SrCl2
Formula Weight: 158.53
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 3.052
Refractive Index: 1.650
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 873°
Boiling Point: 1250°
Merck Reference: 14,8840
MDL No.: MFCD00011249
Formula:
Cl₂Sr MW: 158.53 Boiling Pt: 1250 Melting Pt: 868 Density: 3.052 Storage Temperature: Ambient |
MDL Number:
MFCD00011249 CAS Number: 10476-85-4 EINECS: 233-971-6 Merck Index: 14,8840 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...