Antimony(V) chloride, ultra dry ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
035677-03
AA35677-03EA
86.8
USD
AA35677-03
Antimony(V) chloride, ultra dry ≥99.999% (metals basis)
Antimony (V) chloride
MDL: MFCD00011213
Fieser: 1,42 4,21 5,19 6,22 7,12 15,14 17,17 20,16
Purity: 99.999%
CAS No.: 7647-18-9
Molecular Formula: SbCl5
Formula Weight: 299.02
Physical Form: Liquid, Ampouled under argon
Density: 2.336
Refractive Index: 1.601
Sensitivity: Moisture Sensitive
Odor: Pungent
Melting Point: 2.8°
Boiling Point: 79°/22mm
Merck Reference: 14,695
UN No.: 1730
MDL No.: MFCD00011213
Formula:
Cl₅Sb Boiling Pt: 79 °C (22 mmHg) Melting Pt: 2.8 °C Density: 2.336 Storage Temperature: Ambient |
MDL Number:
MFCD00011213 CAS Number: 7647-18-9 EINECS: 231-601-8 UN: 1730 ADR: 8,II Merck Index: 14,695 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...