Sulfuric acid standardised solution 0.5 N in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
035653-K2
035653-M4
035653-K7
AA35653-K2EA
74.59
USD
AA35653-K2
AA35653-M4
AA35653-K7
Sulfuric acid standardised solution 0.5 N in aqueous solution
Sulfuric acid
MDL: MFCD00064589
Molecular Formula: H2SO4
Formula Weight: 98.08
Physical Form: Liquid
Odor: Odorless
UN No.: 2796
MDL No.: MFCD00064589
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 3264 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...