Calcium chloride 2,75% (w/v) in aqueous solution
Supplier: Thermo Scientific Chemicals
Synonyms:
Chloro calcium, Calcium dichloride, CaCl2
035652-K2
035652-AP
AA35652-K2EA
50.04
USD
AA35652-K2
AA35652-AP
Calcium chloride 2,75% (w/v) in aqueous solution
Calcium chloride
Molecular Formula: CaCl2
Formula Weight: 110.99
Physical Form: Liquid
MDL No.: MFCD00010903
Formula:
CaCl₂ Storage Temperature: Ambient |
MDL Number:
MFCD00010903 CAS Number: 10043-52-4 EINECS: 233-140-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...