Sodium hydroxide standardised solution 0.05 N in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

035623-K7 035623-K2 035623-M4
AA35623-K7EA 89.43 USD
AA35623-K7 AA35623-K2 AA35623-M4
Sodium hydroxide standardised solution 0.05 N in aqueous solution
Sodium hydroxide

MDL: MFCD00003548


CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Odor: Odorless MDL No.: MFCD00003548

Formula: NaOH
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR