Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution volumetric solution

Supplier: Thermo Scientific Chemicals

035622-K2
AA35622-K2EA 108.91 USD
AA35622-K2
Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution volumetric solution
Oxalic acid

MDL: MFCD00002573


Molecular Formula: HO2CCO2H Formula Weight: 90.04 Physical Form: Liquid Appearance: Colorless Odor: Odorless MDL No.: MFCD00002573

Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR