Sodium perborate tetrahydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

033383-36 033383-14 033383-22
AA33383-36EA 98.4 USD
AA33383-36 AA33383-14 AA33383-22
Sodium perborate tetrahydrate, REacton®, Reagent Grade

Bleaching agent for domestic detergents and industrial laundries


CAS No.: 10486-00-7 Molecular Formula: NaBO3.4H2O Formula Weight: 153.86 (81.84anhy) Physical Form: Granular Density: 1.73 Sensitivity: Hygroscopic Appearance: White Odor: Odorless pH: ca 10 Melting Point: 60° dec. Merck Reference: 14,8652 UN No.: 1479 MDL No.: MFCD00149231


Bleaching agent for domestic detergents and industrial laundries

Formula: NaH₂BO₄·4H₂O
Melting Pt: 60 °C (dec.)
Density: 1.73
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9
UN: 1479
ADR: 5.1,III
Merck Index: 14,8652

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR