Sodium perborate tetrahydrate, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
Bleaching agent for domestic detergents and industrial laundries
CAS No.: 10486-00-7
Molecular Formula: NaBO3.4H2O
Formula Weight: 153.86 (81.84anhy)
Physical Form: Granular
Density: 1.73
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
pH: ca 10
Melting Point: 60° dec.
Merck Reference: 14,8652
UN No.: 1479
MDL No.: MFCD00149231
Bleaching agent for domestic detergents and industrial laundries
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...