Sodium diethyldithiocarbamate trihydrate ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00150617
Beilstein Registry No.: 3920507
CAS No.: 20624-25-3
Molecular Formula: (C2H5)2NCS2Na.3H2O
Formula Weight: 225.31 (171.28anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White to off-white
Melting Point: 94-102°
Beilstein Reference: 3920507
Merck Reference: 14,3378
MDL No.: MFCD00150617
Formula:
(C₂H₅)₂NCSSNa·3H₂O Melting Pt: 94…102 °C Density: 1.1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150617 CAS Number: 20624-25-3 EINECS: 205-710-6 UN: 3077 ADR: 9,III Merck Index: 14,3378 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...