Potassium hexacyanoferrate(III) ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Warning
|
MDL: MFCD00011392
Fieser: 1,929 2,345 4,406 5,554 6,480 7,300 8,410 9,385 13,255
Purity: 99.0% min
CAS No.: 13746-66-2
Molecular Formula: K3Fe(CN)6
Formula Weight: 329.26
Physical Form: Crystalline
Density: 1.89
Melting Point: dec.
Merck Reference: 14,7630
MDL No.: MFCD00011392
Formula:
K₃Fe(CN)₆ Melting Pt: Dec. Density: 1.89 Storage Temperature: Ambient |
MDL Number:
MFCD00011392 CAS Number: 13746-66-2 EINECS: 237-323-3 Merck Index: 14,7630 |
Specification Test Results
Insoluble matter | 0.005% |
Cl | 0.01% |
SO3 | 0.01% |
Ferro compounds | 0.05% as ferrocyanide radical |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...