Manganese(II) sulfate monohydrate 98.0-101.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

033341-A3 033341-36
AA33341-A3EA 157 USD
AA33341-A3 AA33341-36
Manganese(II) sulfate monohydrate 98.0-101.0% ACS
Manganese (II) sulphate monohydrate

MDL: MFCD00149159 Notes: Loses all water at 400-450°C


Purity: 98.0-101.0% CAS No.: 10034-96-5 Molecular Formula: MnSO4.H2O Formula Weight: 169.02 (151.00anhy) Physical Form: Powder Density: 2.95 pH: 3-3.5 Merck Reference: 14,5739 UN No.: 3077 MDL No.: MFCD00149159

Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Density: 2.95
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 14,5739

Order Now

Specification Test Results

Insoluble matter 0.01%
Loss on ignition 10.0-12.0
Ca 0.005%
Cl 0.005%
Heavy Metals (as Pb) 0.002%
Fe 0.002%
MG 0.00005
Ni 0.0002
K 0.01%%
NA 0.05%%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR