Magnesium sulfate, anhydrous ≥99.5%

Supplier: Thermo Scientific Chemicals

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

033337-36 033337-A3 033337-A9
AA33337-36EA 98.47 USD
AA33337-36 AA33337-A3 AA33337-A9
Magnesium sulfate, anhydrous ≥99.5%
Magnesium sulphate

Suitable for sensitive molecular biology applications


Purity: 99.5% min CAS No.: 7487-88-9 Molecular Formula: MgSO4 Formula Weight: 120.37 Physical Form: Powder Density: 2.66 Appearance: White Odor: Odorless Melting Point: 1124° dec. Merck Reference: 14,5691 MDL No.: MFCD00011110


Suitable for sensitive molecular biology applications

Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1127 (dec.)
Density: 2.66
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 14,5691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR