Iron(III) nitrate nonahydrate 98.0-101.0% ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

033315-A3 033315-36 033315-F7
AA33315-A3EA 258.44 USD
AA33315-A3 AA33315-36 AA33315-F7
Iron(III) nitrate nonahydrate 98.0-101.0% ACS
Iron (III) nitrate nonahydrate

MDL: MFCD00149708 Freely soluble in water, alcohol, acetone. Slightly soluble in cold and concentrated HNO3


Purity: 98.0-101.0% CAS No.: 7782-61-8 Molecular Formula: FeN3O9.9H2O Formula Weight: 403.99 (241.86anhy) Physical Form: Crystalline Density: 1.684 Appearance: Pale purple Odor: Acidic Melting Point: 47° Boiling Point: 125° dec. Solubility: Freely soluble in water, alcohol, acetone. Slightly soluble in cold and concentrated HNO3 Merck Reference: 14,4027 UN No.: 1466 MDL No.: MFCD00149708

Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125 (dec.)
Melting Pt: 47
Density: 1.684
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1 III
Merck Index: 14,4027

Order Now

Specification Test Results

Insoluble matter 0.005%
Cl 5 ppm
SO3 0.01%
Ca 0.01%
MG 0.005%
K 0.005%
NA 0.0005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR