Cyclohexanone 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pimelic Ketone
033309-AP
033309-K7
033309-M1
033309-K2
AA33309-APEA
70.46
USD
AA33309-AP
AA33309-K7
AA33309-M1
AA33309-K2
Cyclohexanone 99+% ACS
Cyclohexanone
MDL: MFCD00001625
Beilstein Registry No.: 385735
Fieser: 1,178
Purity: 99+%
CAS No.: 108-94-1
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 47°(116°F)
Physical Form: Liquid
Density: 0.9478
Refractive Index: 1.4507
Melting Point: -26°
Boiling Point: 154-156°
Beilstein Reference: 385735
Merck Reference: 14,2726
UN No.: 1915
MDL No.: MFCD00001625
Formula:
C₆H₁₀(=O) MW: 98.14 g/mol Boiling Pt: 155.6 Melting Pt: -31 Density: 0.9478 Flash Pt: 47 °C (116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001625 CAS Number: 108-94-1 EINECS: 203-631-1 UN: 1915 ADR: 3 III REACH: 01-2119453616-35 Merck Index: 14,2726 |
|
Specification Test Results
Color (APHA) | 10 |
Evaporation residue | 0.0005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...