Copper(II) carbonate basic Cu 55%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)

033305-A3 033305-36
AA33305-A3EA 213 USD
AA33305-A3 AA33305-36
Copper(II) carbonate basic Cu 55%
Copper (II) carbonate basic

MDL: MFCD00010976


CAS No.: 12069-69-1 Molecular Formula: CH2Cu2O5 Formula Weight: 221.11 Physical Form: Powder Density: 4.00 Appearance: Green Melting Point: 200° dec. Merck Reference: 14,2631 MDL No.: MFCD00010976

Formula: CuCO₃·Cu(OH)₂
Melting Pt: 200 °C (dec.)
Density: 4.00
Storage Temperature: Ambient
MDL Number: MFCD00010976
CAS Number: 12069-69-1
EINECS: 235-113-6
UN: 3077
ADR: 9,III
Merck Index: 14,2631

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR