Charcoal activated, powder (total ash max 4%)

Supplier: Thermo Scientific Chemicals
Danger

033302-22 033302-36
AA33302-22EA 46.21 USD
AA33302-22 AAA33302-36
Charcoal activated, powder (total ash max 4%)
Charcoal activated

MDL: MFCD00133992


CAS No.: 7440-44-0 Appearance: Black UN No.: 1362 MDL No.: MFCD00133992

Formula: C
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR