Calcium fluoride 97%, REacton®, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
033297-36
033297-22
AA33297-36EA
71.92
USD
AA33297-36
AA33297-22
Calcium fluoride 97%, REacton®, Reagent Grade
Calcium fluoride
Spectroscopy, electronics, laser, high temperature dry-film lubricants
Purity: 97%
CAS No.: 7789-75-5
Molecular Formula: CaF2
Formula Weight: 78.08
Physical Form: Powder
Density: 3.18
Refractive Index: 1.434
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 1403°
Boiling Point: ca 2500°
Solubility: Insoluble in water. Soluble in ammonium salts. Soluble in conc. mineral acids with evolution of HF
Merck Reference: 14,1667
MDL No.: MFCD00010907
Spectroscopy, electronics, laser, high temperature dry-film lubricants
Formula:
CaF₂ MW: 78.07 g/mol Boiling Pt: 2500 Melting Pt: 1418 Density: 3.18 Storage Temperature: Ambient |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 Merck Index: 14,1667 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...