Amidosulfonic acid 99.3-100.3% (dried basis) ACS

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Sulfamic acid, Sulfamidic acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid, Aminosulfonic acid, Amidosulfonic acid, Amidosulfuric Acid, Amidosulfuric acid

033233-22 033233-36
AA33233-22EA 68.18 USD
AA33233-22 AA33233-36
Amidosulfonic acid 99.3-100.3% (dried basis) ACS
Amidosulphonic acid

MDL: MFCD00011603 Fieser: 1,1117 20,353


Purity: 99.3-100.3% CAS No.: 5329-14-6 Molecular Formula: H2NSO3H Formula Weight: 97.09 Physical Form: Crystalline Density: 2.12 pH: 1.2 Melting Point: ca 205° dec. Merck Reference: 14,8921 UN No.: 2967 MDL No.: MFCD00011603

Formula: H₃NO₃S
MW: 97.09 g/mol
Boiling Pt: 247
Melting Pt: 205 (dec.)
Density: 2.12
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8 III
Merck Index: 14,8921
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble matter 0.01%
Residue after ignition 0.01%
Cl 0.001%
SO3 0.0005
Heavy Metals (as Pb) 0.001%
Fe 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR